(R)-(-)-1-Methoxy-2-propanol 96%

Supplier: Ambeed
Danger

A151948-10G A151948-25G A151948-5G
77137-800EA 20.6 USD
77137-800 77137-802 77137-804
(R)-(-)-1-Methoxy-2-propanol 96%
(R)-(-)-1-Methoxy-2-propanol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MDL Number: MFCD01632587
CAS Number: 4984-22-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR