(R)-3-Amino-3-(3-(trifluoromethyl)phenyl)propanoic acid 96%

Supplier: Ambeed
Warning

A151886-100MG A151886-250MG A151886-5G
77137-794EA 38.2 USD
77137-794 77137-796 77137-798
(R)-3-Amino-3-(3-(trifluoromethyl)phenyl)propanoic acid 96%
(R)-3-Amino-3-(3-(trifluoromethyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
MDL Number: MFCD04113664
CAS Number: 793663-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR