trans-4-Methoxycinnamaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(4-Methoxyphenyl)acrylaldehyde

A151746-1G A151746-5G
77137-792EA 7.43 USD
77137-792 77567-190
trans-4-Methoxycinnamaldehyde 95%
trans-4-Methoxycinnamaldehyde

Precaución: For research use only.

Formula: C₁₀H₁₀O₂
MW: 162.19
MDL Number: MFCD00017343
CAS Number: 24680-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR