CGP-57380 98+%

Supplier: Ambeed
Warning

Synonyms: N3-(4-Fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidine-3,4-diamine, CGP 57380, N3-(4-Fluorophenyl)-1H-pyrazolo-[3,4-d]pyrimidine-3,4-diamine

A151588-10MG A151588-1MG A151588-50MG A151588-5MG
77137-784EA 88.06 USD
77137-784 77137-786 77137-788 77137-790
CGP-57380 98+%
CGP-57380

Precaución: for Research Use Only

Formula: C₁₁H₉FN₆
MW: 244.23 g/mol
MDL Number: MFCD03861062
CAS Number: 522629-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR