Triphenylphosphine oxide 97%

Supplier: Ambeed
Warning

A151260-100G A151260-25G A151260-500G A151260-1000G
77137-778EA 7.21 USD
77137-778 77137-780 77137-782 77685-257
Triphenylphosphine oxide 97%
Triphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₈H₁₅OP
MW: 278.29 g/mol
Flash Pt: 180
MDL Number: MFCD00002080
CAS Number: 791-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR