(R)-2-(Piperidin-2-yl)ethanol hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A151157-250MG
A151157-1G
77137-776EA
76.39
USD
77137-776
77567-182
(R)-2-(Piperidin-2-yl)ethanol hydrochloride 97%
(R)-2-(Piperidin-2-yl)ethanol hydrochloride
Precaución: For research use only.
Formula:
C₇H₁₆ClNO MW: 165.66 g/mol |
MDL Number:
MFCD09265047 CAS Number: 787622-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...