2-Cyano-5-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxypyridine-2-carbonitrile
A150764-1G
A150764-250MG
A150764-5G
77137-760EA
58.35
USD
77137-760
77137-762
77137-764
2-Cyano-5-hydroxypyridine 98%
2-Cyano-5-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₆H₄N₂O MW: 120.1100 g/mol |
MDL Number:
MFCD07368197 CAS Number: 86869-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...