(S)-(2-Chloro-5-iodophenyl)(4-((tetrahydrofuran-3-yl)oxy)phenyl)methanone 97%

Supplier: Ambeed
Warning

A150445-10G A150445-1G A150445-250MG A150445-5G
77137-748EA 51.99 USD
77137-748 77137-750 77137-752 77615-216
(S)-(2-Chloro-5-iodophenyl)(4-((tetrahydrofuran-3-yl)oxy)phenyl)methanone 97%
(S)-(2-Chloro-5-iodophenyl)(4-((tetrahydrofuran-3-yl)oxy)phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₇H₁₄ClIO₃
MW: 428.64 g/mol
MDL Number: MFCD22665921
CAS Number: 915095-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR