1-(4-Hydroxy-3,5-dimethoxyphenyl)propan-2-one 95%

Supplier: Ambeed
Warning

A150217-100MG A150217-1G A150217-250MG
77137-722EA 111.23 USD
77137-722 77137-724 77137-726
1-(4-Hydroxy-3,5-dimethoxyphenyl)propan-2-one 95%
1-(4-Hydroxy-3,5-dimethoxyphenyl)propan-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MDL Number: MFCD00210412
CAS Number: 19037-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR