(R)-5-Bromomethyl-2-pyrrolidinone 97%

Supplier: Ambeed
Warning

Synonyms: (R)-5-(Bromomethyl)pyrrolidin-2-one

A149809-100MG A149809-10G A149809-1G A149809-250MG A149809-5G
77137-712EA 9.55 USD
77137-712 77137-714 77137-716 77137-718 77137-720
(R)-5-Bromomethyl-2-pyrrolidinone 97%
(R)-5-Bromomethyl-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₅H₈BrNO
MW: 178.027 g/mol
MDL Number: MFCD06799465
CAS Number: 98612-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR