(Propane-2,2-diylbis(2-hydroxybenzene-5,3,1-triyl))tetramethanol 97%

Supplier: Ambeed
Warning

A149594-10G A149594-1G A149594-250MG A149594-5G
77137-702EA 148.32 USD
77137-702 77137-704 77137-706 77137-708
(Propane-2,2-diylbis(2-hydroxybenzene-5,3,1-triyl))tetramethanol 97%
(Propane-2,2-diylbis(2-hydroxybenzene-5,3,1-triyl))tetramethanol

Precaución: for Research Use Only

Formula: C₁₉H₂₄O₆
MDL Number: MFCD00071732
CAS Number: 3957-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR