2-Chloro-4-morpholinopyrimidine-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A149379-100MG A149379-1G A149379-250MG
77137-694EA 30.77 USD
77137-694 77137-696 77137-698
2-Chloro-4-morpholinopyrimidine-5-carbaldehyde 95%
2-Chloro-4-morpholinopyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀ClN₃O₂
MW: 227.64 g/mol
MDL Number: MFCD27991350
CAS Number: 1820740-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR