Tributyloctylphosphonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Tributyl(octyl)phosphonium bromide

A148364-100G A148364-25G A148364-5G
77137-672EA 152.43 USD
77137-672 77137-674 77137-676
Tributyloctylphosphonium bromide 98%
Tributyloctylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₀H₄₄BrP
MDL Number: MFCD04038147
CAS Number: 57702-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR