Methyl 8-hydroxyquinoline-4-carboxylate 97%

Supplier: Ambeed
Warning

A148102-100MG A148102-250MG
77137-660EA 81.69 USD
77137-660 77137-662
Methyl 8-hydroxyquinoline-4-carboxylate 97%
Methyl 8-hydroxyquinoline-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.19 g/mol
MDL Number: MFCD18417607
CAS Number: 71294-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR