(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 97%

Supplier: Ambeed
Warning

Synonyms: (1S)-1-(2-chlorophenyl)ethane-1,2-diol

A147904-1G A147904-25G
77137-644EA 21.22 USD
77137-644 77137-646
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 97%
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.608 g/mol
MDL Number: MFCD01863241
CAS Number: 133082-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR