(5-Fluoro-2-nitrophenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Fluoro-2-nitrophenylacetic acid
A147637-100G
A147637-10G
A147637-1G
A147637-250MG
A147637-25G
A147637-5G
77137-612EA
639.73
USD
77137-612
77137-614
77137-616
77137-618
77137-620
77137-622
(5-Fluoro-2-nitrophenyl)acetic acid 97%
(5-Fluoro-2-nitrophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₆FNO₄ MW: 199.14 g/mol |
MDL Number:
MFCD07787601 CAS Number: 29640-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...