5-Chloro-2-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-methylnitrobenzene

A147550-10G A147550-25G A147550-500G
77137-604EA 6.18 USD
77137-604 77137-606 77137-608 77137-610
5-Chloro-2-nitrotoluene 98%
5-Chloro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD00024579
CAS Number: 5367-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR