1,2-Dibromo-4-chlorobenzene 97%

Supplier: Ambeed
Warning

A146950-1G A146950-5G A146950-25G A146950-10G
77137-590EA 17.51 USD
77137-590 77137-592 77615-174 77685-238
1,2-Dibromo-4-chlorobenzene 97%
1,2-Dibromo-4-chlorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂Cl
MDL Number: MFCD00672970
CAS Number: 60956-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR