2-Bromo-8-fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 95%

Supplier: Ambeed
Warning

A146434-250MG A146434-1G A146434-5G
77137-560EA 41.21 USD
77137-560 77592-740 77685-237
2-Bromo-8-fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 95%
2-Bromo-8-fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one

Precaución: for Research Use Only

Formula: C₁₁H₈BrFN₂O
MW: 283.10 g/mol
MDL Number: MFCD23703297
CAS Number: 283173-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR