(4S,5R)-3-Benzoyl-2-(4-methoxyphenyl)-4-phenyloxazolidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A146106-1G A146106-250MG
77137-546EA 28.65 USD
77137-546 77137-548
(4S,5R)-3-Benzoyl-2-(4-methoxyphenyl)-4-phenyloxazolidine-5-carboxylic acid 97%
(4S,5R)-3-Benzoyl-2-(4-methoxyphenyl)-4-phenyloxazolidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₅
MW: 403.43 g/mol
MDL Number: MFCD11617943
CAS Number: 949023-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR