3-Bromo-2,4-difluoro-1,5-dimethoxybenzene 97%

Supplier: Ambeed
Warning

A146029-1G A146029-250MG A146029-5G
77137-540EA 65.78 USD
77137-540 77137-542 77137-544
3-Bromo-2,4-difluoro-1,5-dimethoxybenzene 97%
3-Bromo-2,4-difluoro-1,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O₂
MW: 253.04 g/mol
MDL Number: MFCD24539372
CAS Number: 1700265-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR