2-Methylimidazo[1,2-a]pyridin-6-amine 97%

Supplier: Ambeed
Warning

A146000-100MG A146000-250MG A146000-1G
77137-536EA 15.45 USD
77137-536 77137-538 77549-782
2-Methylimidazo[1,2-a]pyridin-6-amine 97%
2-Methylimidazo[1,2-a]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD09991758
CAS Number: 860258-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR