7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine 96%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-N-(p-tolyl)thiazolo[5,4-d]pyrimidin-2-amine

A145766-100MG A145766-1G A145766-250MG
77137-530EA 23.34 USD
77137-530 77137-532 77137-534
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine 96%
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₄S
MW: 276.745 g/mol
MDL Number: MFCD08437191
CAS Number: 871266-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR