4-Chloro-5-fluoro-2-iodoaniline 97%

Supplier: Ambeed
Warning

A145581-100MG A145581-250MG
77137-526EA 27.81 USD
77137-526 77137-528
4-Chloro-5-fluoro-2-iodoaniline 97%
4-Chloro-5-fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄ClFIN
MDL Number: MFCD26407029
CAS Number: 1094759-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR