CPI-455 98%

Supplier: Ambeed
Warning

Synonyms: 6-Isopropyl-7-oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carbonitrile

A145136-10MG A145136-1MG A145136-250MG A145136-5MG A145136-1G
77137-504EA 19.1 USD
77137-504 77137-506 77137-508 77137-510 77685-232
CPI-455 98%
CPI-455

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₄O
MW: 278.31 g/mol
MDL Number: MFCD29921343
CAS Number: 1628208-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR