YK-4-279 98%

Supplier: Ambeed
Warning

Synonyms: 4,7-dichloro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-1H-indol-2-one, 4,7-dichloro-3-hydroxy-3-(2-(4-methoxyphenyl)-2-oxoethyl)indolin-2-one, 4,7-Dichloro-1,3-dihydro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-2H-indol-2-one

A144944-10MG A144944-1MG A144944-50MG A144944-5MG
77137-496EA 20.16 USD
77137-496 77137-498 77137-500 77137-502
YK-4-279 98%
YK-4-279

Precaución: for Research Use Only

Formula: C₁₇H₁₃Cl₂NO₄
MW: 366.20 g/mol
MDL Number: MFCD18382120
CAS Number: 1037184-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR