2-(Trifluoromethyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid

A144870-100G A144870-10G A144870-1G A144870-25G A144870-500G A144870-5G
77137-484EA 126.25 USD
77137-484 77137-486 77137-488 77137-490 77137-492 77137-494
2-(Trifluoromethyl)phenylboronic acid 95%
2-(Trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00236059
CAS Number: 1423-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR