3-Amino-5-methoxypyridine 95%

Supplier: Ambeed
Warning

A144309-10G A144309-1G A144309-250MG A144309-5G A144309-25G
77137-470EA 90.64 USD
77137-470 77137-472 77137-474 77137-476 77685-226
3-Amino-5-methoxypyridine 95%
3-Amino-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD09701369
CAS Number: 64436-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR