3,4-Difluorobenzamide 98%

Supplier: Ambeed
Warning

A144051-1G A144051-25G A144051-5G
77137-452EA 10.3 USD
77137-452 77137-454 77137-456
3,4-Difluorobenzamide 98%
3,4-Difluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅F₂NO
MDL Number: MFCD00015549
CAS Number: 85118-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR