Benzyl (piperidin-4-ylmethyl)carbamate 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (Cbz-4-aminomethyl)piperidine

A143855-1G A143855-25G A143855-5G
77137-450EA 66.96 USD
77137-450 77746-011 77746-012
Benzyl (piperidin-4-ylmethyl)carbamate 98% (by HPLC)
Benzyl (piperidin-4-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.32 g/mol
MDL Number: MFCD07772080
CAS Number: 132431-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR