(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 97%

Supplier: Ambeed
Warning

Synonyms: Rasagiline, (1R)-N-prop-2-ynylindan-1-amine

A143130-100MG A143130-1G A143130-250MG A143130-5G
77137-412EA 11.67 USD
77137-412 77137-414 77137-416 77137-418
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 97%
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.2383 g/mol
MDL Number: MFCD00866571
CAS Number: 136236-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR