2-Methyl-4-(methylthio)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-(methylsulphanyl)benzoic acid

A142419-1G A142419-250MG A142419-5G
77137-374EA 41.38 USD
77137-374 77137-376 77137-378
2-Methyl-4-(methylthio)benzoic acid 97%
2-Methyl-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
MW: 182.24 g/mol
MDL Number: MFCD20651197
CAS Number: 118939-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR