(R)-(-)-2-Chloro-1-propanol 95%

Supplier: Ambeed
Danger

A142004-100MG A142004-1G A142004-250MG A142004-25G A142004-5G
77137-348EA 7.21 USD
77137-348 77137-350 77137-352 77137-354 77137-356
(R)-(-)-2-Chloro-1-propanol 95%
(R)-(-)-2-Chloro-1-propanol

Precaución: for Research Use Only

Formula: H₃CCHClCH₂OH
MDL Number: MFCD00067035
CAS Number: 37493-14-4
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR