3-(Aminocarbonyl)-5-nitrobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carbamoyl-5-nitrobenzeneboronic acid, (3-Carbamoyl-5-nitrophenyl)boronic acid

A141833-1G A141833-250MG A141833-5G
77137-338EA 122.01 USD
77137-338 77137-340 77137-342
3-(Aminocarbonyl)-5-nitrobenzeneboronic acid 98%
3-(Aminocarbonyl)-5-nitrobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₅
MW: 209.952 g/mol
MDL Number: MFCD07783870
CAS Number: 102170-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR