(R)-(-)-1-Benzyloxy-2-propanol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1-(Benzyloxy)propan-2-ol
A140836-10G
A140836-1G
A140836-250MG
A140836-5G
A140836-100G
A140836-25G
77137-298EA
207.94
USD
77137-298
77137-300
77137-302
77137-304
77745-997
77745-998
(R)-(-)-1-Benzyloxy-2-propanol 95%
(R)-(-)-1-Benzyloxy-2-propanol
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₂ MW: 166.217 g/mol |
MDL Number:
MFCD07367007 CAS Number: 89401-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...