(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A140815-100MG
A140815-1G
A140815-250MG
A140815-25G
77137-290EA
9.27
USD
77137-290
77137-292
77137-294
77137-296
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid 95%
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₅ |
MDL Number:
MFCD09260604 CAS Number: 884512-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...