Apilimod mesylate 98%

Supplier: Ambeed
Warning

Synonyms: (E)-4-(6-(2-(3-methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine dimethanesulphonate, STA 5326 mesylate, 4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine dimethanesulphonate, Apilimod dimesylate

A140672-1MG A140672-5MG A140672-100MG
77137-282EA 21.22 USD
77137-282 77137-284 77745-993
Apilimod mesylate 98%
Apilimod mesylate

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₆O₈S₂
MW: 610.70 g/mol
MDL Number: MFCD28167749
CAS Number: 870087-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR