2-Chloroquinazoline 95%

Supplier: Ambeed
Warning

A140401-1G A140401-250MG A140401-5G
77137-270EA 50.93 USD
77137-270 77137-272 77137-274
2-Chloroquinazoline 95%
2-Chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₅ClN₂
MW: 164.5900 g/mol
MDL Number: MFCD09959715
CAS Number: 6141-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR