(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1S,3R)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-FMOC protected, (1S,3R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopentanecarboxylic acid

A139908-250MG A139908-100MG
77137-266EA 44.56 USD
77137-266 77685-197
(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid 95%
(1S,3R)-Fmoc-3-aminocyclopentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD01311757
CAS Number: 220497-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR