(2-Fluorophenyl)glycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-(2-fluorophenyl)acetic acid

A138651-1G A138651-250MG
77137-230EA 23.69 USD
77137-230 77137-232
(2-Fluorophenyl)glycine 98%
(2-Fluorophenyl)glycine

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
MDL Number: MFCD00042726
CAS Number: 84145-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR