3-Chloro-4-methylanisole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methoxy-1-methylbenzene, 3-Chloro-4-methylanisole, 2-Chloro-4-methoxytoluene

A138587-10G A138587-1G A138587-5G
77137-224EA 22.28 USD
77137-224 77137-226 77137-228
3-Chloro-4-methylanisole 98%
3-Chloro-4-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00087376
CAS Number: 54788-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR