tert-Butyl (3-iodocyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

A138425-100MG A138425-1G A138425-250MG A138425-5G
77137-216EA 15.45 USD
77137-216 77137-218 77137-220 77137-222
tert-Butyl (3-iodocyclobutyl)carbamate 97%
tert-Butyl (3-iodocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆INO₂
MDL Number: MFCD24530658
CAS Number: 1824239-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR