PD-168393 99+%

Supplier: Ambeed
Warning

Synonyms: PD 168,393, 4-[(3-Bromophenyl)amino]-6-acrylamidoquinazoline, N-[4-[(3-bromophenyl)amino]-6-quinazolinyl]-2-propenamide, N-(4-((3-Bromophenyl)amino)quinazolin-6-yl)acrylamide, PD168393

A137778-100MG A137778-10MG A137778-1MG A137778-25MG A137778-2MG A137778-50MG A137778-5MG
77137-190EA 206.88 USD
77137-190 77137-192 77137-194 77137-196 77137-198 77137-200 77137-202
PD-168393 99+%
PD-168393

Precaución: for Research Use Only

Formula: C₁₇H₁₃BrN₄O
MW: 369.2 g/mol
MDL Number: MFCD02179207
CAS Number: 194423-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR