(S)-(-)-1-(2-Naphthyl)ethylamine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(Naphthalen-2-yl)ethanamine

A137171-100G A137171-10G A137171-1G A137171-250MG A137171-25G A137171-5G
77137-154EA 744.76 USD
77137-154 77137-156 77137-158 77137-160 77137-162 77137-164
(S)-(-)-1-(2-Naphthyl)ethylamine 97%
(S)-(-)-1-(2-Naphthyl)ethylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00085366
CAS Number: 3082-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR