5-Chlorobenzoxazole 98%

Supplier: Ambeed
Warning

A137169-10G A137169-1G A137169-250MG A137169-25G A137169-5G
77137-144EA 18.54 USD
77137-144 77137-146 77137-148 77137-150 77137-152
5-Chlorobenzoxazole 98%
5-Chlorobenzoxazole

Precaución: for Research Use Only

Formula: C₇H₄ClNO
MW: 153.57 g/mol
MDL Number: MFCD00216939
CAS Number: 17200-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR