Methyl 4-bromo-1-methyl-1H-indazole-6-carboxylate 98%

Supplier: Ambeed
Warning

A136713-100MG A136713-250MG
77137-132EA 65.78 USD
77137-132 77137-134
Methyl 4-bromo-1-methyl-1H-indazole-6-carboxylate 98%
Methyl 4-bromo-1-methyl-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.09 g/mol
MDL Number: MFCD28501937
CAS Number: 1638759-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR