Methyl-4-fluoro-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-3-nitrobenzoate, 4-Fluorobenzoic acid methyl ester

A136431-100G A136431-10G A136431-25G A136431-500G A136431-5G
77137-120EA 67.9 USD
77137-120 77137-122 77137-124 77137-126 77137-128
Methyl-4-fluoro-3-nitrobenzoate 98%
Methyl-4-fluoro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
MDL Number: MFCD00196161
CAS Number: 329-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR