5-Bromo-N,N,N-trimethylpentan-1-aminium bromide 98%
Supplier: Ambeed
|
Warning
|
A136364-1G
A136364-25G
A136364-5G
77137-114EA
32.96
USD
77137-114
77137-116
77137-118
5-Bromo-N,N,N-trimethylpentan-1-aminium bromide 98%
5-Bromo-N,N,N-trimethylpentan-1-aminium bromide
Precaución: for Research Use Only
Formula:
C₈H₁₉Br₂N MW: 289.05 g/mol |
MDL Number:
MFCD00545102 CAS Number: 15008-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...