STK-321130 98+%

Supplier: Ambeed
Warning

Synonyms: 5-[(5-Chloro-1H-pyrrolo[2,3-b]pyridin-3-yl)methyl]-N-[[4-(trifluoromethyl)phenyl]methyl]-2-pyridinamine, 5-((5-Chloro-1H-pyrrolo[2,3-b]pyridin-3-yl)methyl)-N-(4-(trifluoromethyl)benzyl)pyridin-2-amine, FLT3-IN-2

A135706-10MG A135706-1MG A135706-25MG A135706-5MG
77137-072EA 103.97 USD
77137-072 77137-074 77137-076 77137-078
STK-321130 98+%
STK-321130

Precaución: for Research Use Only

Formula: C₂₁H₁₆ClF₃N₄
MW: 416.83 g/mol
MDL Number: MFCD29472260
CAS Number: 923562-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR