(+)-Corey lactone 95%

Supplier: Ambeed
Warning

A135656-10G A135656-1G A135656-250MG A135656-25G A135656-5G
77137-066EA 223.52 USD
77137-066 77137-068 77137-070 77685-169 77685-170
(+)-Corey lactone 95%
(+)-Corey lactone

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MDL Number: MFCD00274285
CAS Number: 76704-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR